siRNA Details
| Gene Symbol: Fktn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139309.4 | NP_647470.1 | 4 | 703 | ||
| GenBank mRNA | AB077383.1 | - | 5 | 887 | ||
| AJ511807.1 | - | 5 | 723 | |||
| AK052357.1 | - | 6 | 861 | |||
| AK085931.1 | - | 4 | 704 | |||
| AK142744.1 | - | 863 | ||||
| AK144743.1 | - | 5 | 732 | |||
| BC017538.1 | - | 5 | 719 | 4506804 | ||

Fktn
Genomic Map