siRNA Details
| Gene Symbol: Vmn1r69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145842.3 | NP_665841.1 | 1 | 872 | ||
| GenBank mRNA | AF394954.1 | - | 1 | 871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r69| Gene Symbol: Vmn1r69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145842.3 | NP_665841.1 | 1 | 872 | ||
| GenBank mRNA | AF394954.1 | - | 1 | 871 | ||