siRNA Details
Gene Symbol: Vmn1r69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145842.3 | NP_665841.1 | 1 | 1613 | ||
GenBank mRNA | AF394954.1 | - | 1 | 1612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145842.3 | NP_665841.1 | 1 | 1613 | ||
GenBank mRNA | AF394954.1 | - | 1 | 1612 |