siRNA Details
| Gene Symbol: Olfr317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011769.2 | NP_001011769.2 | 1 | 700 | ||
| GenBank mRNA | BC147131.1 | - | 1 | 700 | 9056194 | |
| BC147132.1 | - | 1 | 700 | 9056174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr317| Gene Symbol: Olfr317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011769.2 | NP_001011769.2 | 1 | 700 | ||
| GenBank mRNA | BC147131.1 | - | 1 | 700 | 9056194 | |
| BC147132.1 | - | 1 | 700 | 9056174 | ||