siRNA Details
| Gene Symbol: Olfr1419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011775.1 | NP_001011775.1 | 1 | 275 | ||
| GenBank mRNA | BC147200.1 | - | 1 | 366 | 9056244 | |
| BC147201.1 | - | 1 | 366 | 9056264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1419| Gene Symbol: Olfr1419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011775.1 | NP_001011775.1 | 1 | 275 | ||
| GenBank mRNA | BC147200.1 | - | 1 | 366 | 9056244 | |
| BC147201.1 | - | 1 | 366 | 9056264 | ||