siRNA Details
| Gene Symbol: Olfr566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011536.1 | NP_001011536.1 | 1 | 254 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr566| Gene Symbol: Olfr566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011536.1 | NP_001011536.1 | 1 | 254 | ||
Set Membership: |
|