siRNA Details
| Gene Symbol: Olfr1308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207151.1 | NP_997034.1 | 1 | 790 | ||
| GenBank mRNA | BC108949.2 | - | 1 | 789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1308| Gene Symbol: Olfr1308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207151.1 | NP_997034.1 | 1 | 790 | ||
| GenBank mRNA | BC108949.2 | - | 1 | 789 | ||