siRNA Details
| Gene Symbol: Olfr1412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146277.1 | NP_666389.1 | 1 | 470 | ||
| GenBank mRNA | BC125241.1 | - | 1 | 470 | 40044004 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1412| Gene Symbol: Olfr1412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146277.1 | NP_666389.1 | 1 | 470 | ||
| GenBank mRNA | BC125241.1 | - | 1 | 470 | 40044004 | |