siRNA Details
Gene Symbol: Olfr1366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146283.2 | NP_666395.2 | 1 | 720 | ||
GenBank mRNA | BC138101.1 | - | 1 | 718 | 8861120 | |
BC138102.1 | - | 1 | 720 | 8861121 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146283.2 | NP_666395.2 | 1 | 720 | ||
GenBank mRNA | BC138101.1 | - | 1 | 718 | 8861120 | |
BC138102.1 | - | 1 | 720 | 8861121 |