siRNA Details
| Gene Symbol: Olfr893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146336.2 | NP_666448.2 | 1 | 458 | ||
| GenBank mRNA | BC130242.1 | - | 1 | 459 | 40142233 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr893| Gene Symbol: Olfr893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146336.2 | NP_666448.2 | 1 | 458 | ||
| GenBank mRNA | BC130242.1 | - | 1 | 459 | 40142233 | |