siRNA Details
| Gene Symbol: Olfr1093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146366.1 | NP_666478.1 | 1 | 675 | ||
| GenBank mRNA | BC138167.1 | - | 1 | 762 | 8861186 | |
More Information
Other Species Targeted
Set Membership: |
|

Olfr1093
Genomic Map