siRNA Details
| Gene Symbol: Olfr1350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146389.1 | NP_666501.1 | 1 | 286 | ||
| GenBank mRNA | BC104062.1 | - | 1 | 285 | ||
| BC104063.1 | - | 1 | 286 | 40040754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1350| Gene Symbol: Olfr1350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146389.1 | NP_666501.1 | 1 | 286 | ||
| GenBank mRNA | BC104062.1 | - | 1 | 285 | ||
| BC104063.1 | - | 1 | 286 | 40040754 | ||