siRNA Details
| Gene Symbol: Olfr694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146452.2 | NP_666663.2 | 1 | 489 | ||
| GenBank mRNA | BC129796.1 | - | 1 | 489 | 40042951 | |
| BC129797.1 | - | 1 | 489 | 40042952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr694| Gene Symbol: Olfr694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146452.2 | NP_666663.2 | 1 | 489 | ||
| GenBank mRNA | BC129796.1 | - | 1 | 489 | 40042951 | |
| BC129797.1 | - | 1 | 489 | 40042952 | ||