siRNA Details
| Gene Symbol: Olfr1391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146468.1 | NP_666679.1 | 1 | 436 | ||
| GenBank mRNA | BC127961.1 | - | 1 | 440 | 40134837 | |
| BC148323.1 | - | 1 | 440 | 40134755 | ||
| BC150143.1 | - | 1 | 440 | 40134757 | ||

Olfr1391
Genomic Map