siRNA Details
| Gene Symbol: Olfr800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146548.1 | NP_666759.1 | 1 | 550 | ||
| GenBank mRNA | BC134373.1 | - | 1 | 554 | 40134856 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr800| Gene Symbol: Olfr800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146548.1 | NP_666759.1 | 1 | 550 | ||
| GenBank mRNA | BC134373.1 | - | 1 | 554 | 40134856 | |