siRNA Details
| Gene Symbol: Olfr873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146561.1 | NP_666772.1 | 1 | 809 | ||
| GenBank mRNA | BC139140.1 | - | 1 | 892 | 8862162 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr873| Gene Symbol: Olfr873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146561.1 | NP_666772.1 | 1 | 809 | ||
| GenBank mRNA | BC139140.1 | - | 1 | 892 | 8862162 | |