siRNA Details
| Gene Symbol: Olfr1015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146571.2 | NP_666782.2 | 1 | 948 | ||
| GenBank mRNA | BC119114.1 | - | 1 | 948 | 8733863 | |
| BC119142.1 | - | 1 | 948 | 8733891 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1015| Gene Symbol: Olfr1015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146571.2 | NP_666782.2 | 1 | 948 | ||
| GenBank mRNA | BC119114.1 | - | 1 | 948 | 8733863 | |
| BC119142.1 | - | 1 | 948 | 8733891 | ||