siRNA Details
| Gene Symbol: Olfr307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146617.1 | NP_666828.1 | 1 | 239 | ||
| GenBank mRNA | BC132055.1 | - | 1 | 262 | 40130332 | |
| BC132437.1 | - | 1 | 262 | 40130714 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr307| Gene Symbol: Olfr307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146617.1 | NP_666828.1 | 1 | 239 | ||
| GenBank mRNA | BC132055.1 | - | 1 | 262 | 40130332 | |
| BC132437.1 | - | 1 | 262 | 40130714 | ||