siRNA Details
| Gene Symbol: Olfr1489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146635.1 | NP_666846.1 | 1 | 294 | ||
| GenBank mRNA | BC111827.1 | - | 1 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1489| Gene Symbol: Olfr1489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146635.1 | NP_666846.1 | 1 | 294 | ||
| GenBank mRNA | BC111827.1 | - | 1 | 293 | ||