siRNA Details
| Gene Symbol: Olfr1164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146641.2 | NP_666852.2 | 1 | 669 | ||
| GenBank mRNA | BC132589.1 | - | 1 | 669 | 40130866 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1164| Gene Symbol: Olfr1164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146641.2 | NP_666852.2 | 1 | 669 | ||
| GenBank mRNA | BC132589.1 | - | 1 | 669 | 40130866 | |