siRNA Details
| Gene Symbol: Olfr1140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146642.2 | NP_666853.2 | 1 | 681 | ||
| GenBank mRNA | BC120608.1 | - | 1 | 681 | 40129531 | |
| BC120610.1 | - | 1 | 681 | 40129533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1140| Gene Symbol: Olfr1140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146642.2 | NP_666853.2 | 1 | 681 | ||
| GenBank mRNA | BC120608.1 | - | 1 | 681 | 40129531 | |
| BC120610.1 | - | 1 | 681 | 40129533 | ||