siRNA Details
| Gene Symbol: Olfr1135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146660.2 | NP_666871.2 | 1 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1135| Gene Symbol: Olfr1135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146660.2 | NP_666871.2 | 1 | 69 | ||