siRNA Details
| Gene Symbol: Olfr820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146675.1 | NP_666886.1 | 1 | 239 | ||
| GenBank mRNA | BC119398.1 | - | 1 | 301 | 8734147 | |
| BC119400.1 | - | 1 | 301 | 8734149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr820| Gene Symbol: Olfr820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146675.1 | NP_666886.1 | 1 | 239 | ||
| GenBank mRNA | BC119398.1 | - | 1 | 301 | 8734147 | |
| BC119400.1 | - | 1 | 301 | 8734149 | ||