siRNA Details
Gene Symbol: Olfr1444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146702.1 | NP_666913.1 | 1 | 140 | ||
GenBank mRNA | BC119059.1 | - | 1 | 175 | 8733808 | |
BC125619.1 | - | 1 | 175 | 40130134 |
