siRNA Details
Gene Symbol: Olfr1342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146713.1 | NP_666924.1 | 1 | 207 | ||
GenBank mRNA | BC119161.1 | - | 1 | 220 | 8733910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146713.1 | NP_666924.1 | 1 | 207 | ||
GenBank mRNA | BC119161.1 | - | 1 | 220 | 8733910 |