siRNA Details
| Gene Symbol: Olfr1342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146713.1 | NP_666924.1 | 1 | 813 | ||
| GenBank mRNA | BC119161.1 | - | 1 | 826 | 8733910 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1342| Gene Symbol: Olfr1342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146713.1 | NP_666924.1 | 1 | 813 | ||
| GenBank mRNA | BC119161.1 | - | 1 | 826 | 8733910 | |