siRNA Details
| Gene Symbol: Olfr419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146715.2 | NP_666926.2 | 1 | 907 | ||
| GenBank mRNA | BC119050.1 | - | 1 | 907 | 8733799 | |
| BC119052.1 | - | 1 | 907 | 8733801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr419| Gene Symbol: Olfr419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146715.2 | NP_666926.2 | 1 | 907 | ||
| GenBank mRNA | BC119050.1 | - | 1 | 907 | 8733799 | |
| BC119052.1 | - | 1 | 907 | 8733801 | ||