siRNA Details
Gene Symbol: Olfr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146747.1 | NP_666958.1 | 1 | 21 | ||
GenBank mRNA | BC132188.1 | - | 1 | 101 | 40130465 | |
BC132190.1 | - | 1 | 101 | 40130467 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146747.1 | NP_666958.1 | 1 | 21 | ||
GenBank mRNA | BC132188.1 | - | 1 | 101 | 40130465 | |
BC132190.1 | - | 1 | 101 | 40130467 |