siRNA Details
| Gene Symbol: Olfr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146747.1 | NP_666958.1 | 1 | 768 | ||
| GenBank mRNA | BC132188.1 | - | 1 | 848 | 40130465 | |
| BC132190.1 | - | 1 | 848 | 40130467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr146| Gene Symbol: Olfr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146747.1 | NP_666958.1 | 1 | 768 | ||
| GenBank mRNA | BC132188.1 | - | 1 | 848 | 40130465 | |
| BC132190.1 | - | 1 | 848 | 40130467 | ||