siRNA Details
| Gene Symbol: Olfr508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146773.1 | NP_666984.1 | 1 | 476 | ||
| GenBank mRNA | BC130241.1 | - | 1 | 480 | 40142176 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr508| Gene Symbol: Olfr508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146773.1 | NP_666984.1 | 1 | 476 | ||
| GenBank mRNA | BC130241.1 | - | 1 | 480 | 40142176 | |