siRNA Details
| Gene Symbol: Olfr1246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146792.2 | NP_667003.2 | 1 | 133 | ||
| GenBank mRNA | BC138403.1 | - | 1 | 133 | 8861423 | |
| BC138404.1 | - | 1 | 133 | 8861424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1246| Gene Symbol: Olfr1246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146792.2 | NP_667003.2 | 1 | 133 | ||
| GenBank mRNA | BC138403.1 | - | 1 | 133 | 8861423 | |
| BC138404.1 | - | 1 | 133 | 8861424 | ||