siRNA Details
| Gene Symbol: Olfr812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146795.1 | NP_667006.1 | 1 | 125 | ||
| GenBank mRNA | BC132194.1 | - | 1 | 147 | 40130471 | |
| BC132196.1 | - | 1 | 147 | 40130473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr812| Gene Symbol: Olfr812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146795.1 | NP_667006.1 | 1 | 125 | ||
| GenBank mRNA | BC132194.1 | - | 1 | 147 | 40130471 | |
| BC132196.1 | - | 1 | 147 | 40130473 | ||