siRNA Details
Gene Symbol: Olfr109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146835.1 | NP_667046.1 | 1 | 660 | ||
GenBank mRNA | BC130246.1 | - | 1 | 664 | 40142352 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146835.1 | NP_667046.1 | 1 | 660 | ||
GenBank mRNA | BC130246.1 | - | 1 | 664 | 40142352 |