siRNA Details
| Gene Symbol: Olfr109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146835.1 | NP_667046.1 | 1 | 510 | ||
| GenBank mRNA | BC130246.1 | - | 1 | 514 | 40142352 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr109| Gene Symbol: Olfr109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146835.1 | NP_667046.1 | 1 | 510 | ||
| GenBank mRNA | BC130246.1 | - | 1 | 514 | 40142352 | |