siRNA Details
| Gene Symbol: Olfr1090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146847.1 | NP_667058.1 | 1 | 908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1090| Gene Symbol: Olfr1090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146847.1 | NP_667058.1 | 1 | 908 | ||