siRNA Details
| Gene Symbol: Olfr982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146854.1 | NP_667065.1 | 1 | 606 | ||
| GenBank mRNA | BC104306.2 | - | 1 | 605 | 40044025 | |
| BC104307.1 | - | 1 | 606 | 40044030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr982| Gene Symbol: Olfr982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146854.1 | NP_667065.1 | 1 | 606 | ||
| GenBank mRNA | BC104306.2 | - | 1 | 605 | 40044025 | |
| BC104307.1 | - | 1 | 606 | 40044030 | ||