siRNA Details
| Gene Symbol: Olfr371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146859.2 | NP_667070.1 | 1 | 439 | ||
| GenBank mRNA | BC127975.1 | - | 1 | 441 | 40135153 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr371| Gene Symbol: Olfr371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146859.2 | NP_667070.1 | 1 | 439 | ||
| GenBank mRNA | BC127975.1 | - | 1 | 441 | 40135153 | |