siRNA Details
| Gene Symbol: Olfr770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146863.1 | NP_667074.1 | 1 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr770| Gene Symbol: Olfr770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146863.1 | NP_667074.1 | 1 | 413 | ||