siRNA Details
| Gene Symbol: Olfr908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146872.1 | NP_667083.1 | 1 | 781 | ||
| GenBank mRNA | BC112370.1 | - | 1 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr908| Gene Symbol: Olfr908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146872.1 | NP_667083.1 | 1 | 781 | ||
| GenBank mRNA | BC112370.1 | - | 1 | 778 | ||