siRNA Details
| Gene Symbol: Olfr900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146874.1 | NP_667085.1 | 1 | 922 | ||
| GenBank mRNA | BC106986.2 | - | 1 | 922 | 40043276 | |
| Gene Symbol: Olfr898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146871.2 | NP_667082.2 | 1 | 922 | ||

Olfr900
Genomic Map