siRNA Details
| Gene Symbol: Olfr900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146874.1 | NP_667085.1 | 1 | 756 | ||
| GenBank mRNA | BC106986.2 | - | 1 | 756 | 40043276 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr900| Gene Symbol: Olfr900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146874.1 | NP_667085.1 | 1 | 756 | ||
| GenBank mRNA | BC106986.2 | - | 1 | 756 | 40043276 | |