siRNA Details
| Gene Symbol: Olfr1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146887.1 | NP_667098.1 | 1 | 437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1301| Gene Symbol: Olfr1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146887.1 | NP_667098.1 | 1 | 437 | ||