siRNA Details
Gene Symbol: Olfr1336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146915.1 | NP_667126.1 | 1 | 810 | ||
GenBank mRNA | BC104285.1 | - | 1 | 809 | ||
BC104286.1 | - | 1 | 810 | 40042538 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
