siRNA Details
| Gene Symbol: Olfr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146986.1 | NP_667197.1 | 1 | 906 | ||
| GenBank mRNA | BC106817.1 | - | 1 | 905 | ||
| BC128054.1 | - | 1 | 906 | 40043588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr38| Gene Symbol: Olfr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146986.1 | NP_667197.1 | 1 | 906 | ||
| GenBank mRNA | BC106817.1 | - | 1 | 905 | ||
| BC128054.1 | - | 1 | 906 | 40043588 | ||