siRNA Details
| Gene Symbol: Olfr195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146998.1 | NP_667209.1 | 1 | 638 | ||
| GenBank mRNA | BC119110.1 | - | 1 | 661 | 8733859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr195| Gene Symbol: Olfr195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146998.1 | NP_667209.1 | 1 | 638 | ||
| GenBank mRNA | BC119110.1 | - | 1 | 661 | 8733859 | |