siRNA Details
| Gene Symbol: Olfr399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147004.2 | NP_667215.2 | 1 | 441 | ||
| GenBank mRNA | BC120820.1 | - | 1 | 441 | 40129743 | |
| BC120846.1 | - | 1 | 441 | 40129769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr399| Gene Symbol: Olfr399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147004.2 | NP_667215.2 | 1 | 441 | ||
| GenBank mRNA | BC120820.1 | - | 1 | 441 | 40129743 | |
| BC120846.1 | - | 1 | 441 | 40129769 | ||