siRNA Details
Gene Symbol: Olfr711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147035.2 | NP_667246.2 | 3 | 804 | ||
GenBank mRNA | BC132382.1 | - | 1 | 475 | 40130659 | |
BC138433.1 | - | 1 | 475 | 8861453 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147035.2 | NP_667246.2 | 3 | 804 | ||
GenBank mRNA | BC132382.1 | - | 1 | 475 | 40130659 | |
BC138433.1 | - | 1 | 475 | 8861453 |