siRNA Details
Gene Symbol: Olfr659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147050.1 | NP_667261.1 | 1 | 640 | ||
GenBank mRNA | BC148221.1 | - | 1 | 644 | 40136035 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147050.1 | NP_667261.1 | 1 | 640 | ||
GenBank mRNA | BC148221.1 | - | 1 | 644 | 40136035 |