siRNA Details
Gene Symbol: Olfr571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147085.2 | NP_667296.2 | 1 | 790 | ||
GenBank mRNA | BC104118.1 | - | 1 | 783 | 40044090 | |
BC104119.1 | - | 1 | 783 | 40044091 | ||
BC119419.1 | - | 1 | 790 | 40044094 | ||
BC119420.1 | - | 1 | 790 | 40044095 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
