siRNA Details
| Gene Symbol: Olfr571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147085.2 | NP_667296.2 | 1 | 149 | ||
| GenBank mRNA | BC104118.1 | - | 1 | 142 | 40044090 | |
| BC104119.1 | - | 1 | 142 | 40044091 | ||
| BC119419.1 | - | 1 | 149 | 40044094 | ||
| BC119420.1 | - | 1 | 149 | 40044095 | ||

Olfr571
Genomic Map